- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse DLL4 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-970
Mouse DLL4 AccuSignal ELISA Kit
- Natural and recombinant mouse DLL4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S28-P525
Delta like ligand 4 is a protein that in humans is encoded by the DLL4 gene. It is mapped to 15q15.1. This gene is a homolog of the Drosophila delta gene. DLL4 is a transmembrane ligand for Notch receptors that shows restricted expression to endothelial cells (ECs), in particular to arteries and capillaries, and is involved in vascular development, and it also appeared to be a major trigger of Notch receptor activities previously implicated in arterial and vascular development. Mouse DLL4 could activate mouse Notch1 and mouse Notch4. DLL4 acts as a negative regulator of tumor angiogenesis, its blockade results in the striking uncoupling of tumor growth from vessel density, presenting a novel therapeutic approach even for tumors resistant to anti-VEGF therapies. In addition to it, this gene also plays an important role in promoting Th17 effector activity during mycobacterial challenge.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse DLL4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse DLL4 standard solution and each sample be measured in duplicate.