- Primary antibody reactivity:Human
- Description:Human CD320 - 8D6A ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-972
Human CD320 - 8D6A AccuSignal ELISA Kit
- Natural and recombinant human CD320.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S36-Y229
CD320(cluster of differentiation 320), also known as 8D6A or TCBLR, is a human gene. It is mapped to chromosome 19p13.2. This gene encodes the transcobalamin receptor that is expressed at the cell surface. It mediates the cellular uptake of transcobalamin bound cobalamin(vitamin B12), and the expression of CD320 enhanced B-cell proliferation and immunoglobulin secretion. Mutations in this gene are associated with methylmalonic aciduria. It has been found that antagonists of the CD320 signaling pathway may counter the growth of follicular lymphomas or other tumors that metastasize to lymphoid follicles. CD320 is also a specific receptor for uptake of TCN2-bound cobalamin.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CD320 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human CD320 standard solution and each sample be measured in duplicate.