Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse TSLP ELISA Kit, Rockland Immunochemicals
Mouse TSLP ELISA Kit, Rockland Immunochemicals
Catalog # 76235-886
CAS Number:  
Mouse TSLP ELISA Kit, Rockland Immunochemicals
Catalog # 76235-886
Supplier Number:  KOA0703
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    TSLP
  • Description:
    Mouse TSLP ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-886

Specifications

About this item

Mouse TSLP AccuSignal ELISA Kit

  • Natural and recombinant mouse TSLP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Y20-E140

Thymic stromal lymphopoietin, also called TSLP, is a protein belonging to the cytokine family. This gene is mapped to 5q22.1. It encodes a hemopoietic cytokine proposed to signal through a heterodimeric receptor complex composed of the thymic stromal lymphopoietin receptor and the IL-7R alpha chain. It mainly impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes and enhances the maturation of CD11c(+) dendritic cells. The protein promotes T helper type 2(TH2) cell responses that are associated with immunity in various inflammatory diseases, including asthma, allergic inflammation and chronic obstructive pulmonary disease. The protein is therefore considered a potential therapeutic target for the treatment of such diseases.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse TSLP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TSLP standard solution and each sample be measured in duplicate.