ContactUsLinkComponent
Human MFGE8 - Lactadherin ELISA Kit, Rockland Immunochemicals
  76235-876
 :  
Human MFGE8 - Lactadherin ELISA Kit, Rockland Immunochemicals
  76235-876
 :  KOA0698
 :  
  • Primary antibody reactivity:
    Human
  • Description:
    Human MFGE8 - Lactadherin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-876
Human MFGE8 - Lactadherin AccuSignal ELISA Kit
  • Natural and recombinant human MFGE8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: L24-C387
MFGE8(Milk Fat Globule-Egf Factor 8), also called as Lactadherin or SED1, is a protein which in humans is encoded by the MFGE8 gene. Mfge8 is secreted protein found in vertebrates, including mammals as well as birds. By fluorescence in situ hybridization, the MFGE8 gene was mapped to chromosome 15q25. MFGE8 is a factor that links apoptotic cells to phagocytes. It specifically bound to apoptotic cells by recognizing aminophospholipids such as phosphatidylserine. MFGE8, when engaged by phospholipids, bound to cells via its RGD motif. It bound particularly strongly to cells expressing alpha-V-beta-3 integrin. It showed that Mfge8 was expressed in intestinal lamina propria macrophages in mice. Using a wound-healing assay, they showed that Mfge8 promoted migration of intestinal epithelial cells through a PKC-epsilon(PRKCE)-dependent mechanism.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human MFGE8 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva, urine or human milk to each empty well. We recommend that each human MFGE8 standard solution and each sample is measured in duplicate.