ContactUsLinkComponent
Human LIFR ELISA Kit, Rockland Immunochemicals
  76235-874
 :  
Human LIFR ELISA Kit, Rockland Immunochemicals
  76235-874
 :  KOA0697
 :  
  • Primary antibody reactivity:
    Human
  • Description:
    Human LIFR ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-874
Human LIFR AccuSignal ELISA Kit
  • Natural and recombinant human LIFR.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q45-S833
LIFR, also known as CD118(Cluster of Differentiation 118), is a subunit of a receptor for leukemia inhibitory factor. This gene encodes a protein that belongs to the type I cytokine receptor family. It is mapped to 5p31.1. The LIF receptor(LIFR) is the low-affinity binding chain that, together with the high-affinity converter subunit gp130, forms a high-affinity receptor complex that mediates the action of the leukemia-inhibitory factor. LIF is a polyfunctional cytokine that affects the differentiation, survival, and proliferation of a wide variety of cells in the adult and the embryo. Mutations in this gene cause Schwartz-Jampel syndrome type 2, a disease belonging to the group of the bent-bone dysplasias. A translocation that involves the promoter of this gene, togerther with the pleiomorphic adenoma gene 1, is associated with salivary gland pleiomorphic adenoma, a common type of benign epithelial tumor of the salivary gland.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human LIFR standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human LIFR standard solution and each sample be measured in duplicate.