Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse ACE2 ELISA Kit, Rockland Immunochemicals
Mouse ACE2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-854
CAS Number:  
Mouse ACE2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-854
Supplier Number:  KOA0687
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse ACE2 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-854

Specifications

About this item

Mouse ACE2 AccuSignal ELISA Kit

  • Natural and recombinant mouse ACE2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: Q18-T740

AngiotensinI-converting enzyme 2(ACE 2) is a protein belongs to the angiotensin-converting enzyme family of dipeptidyl carboxydipeptidases and has considerable homology to human angiotensin 1 converting enzyme. By sequence similarity to a sequence in GenBank, this gene is mapped to Xp22.2. This secreted protein catalyzes the cleavage of angiotensin I into angiotensin 1-9, and angiotensin II into the vasodilator angiotensin 1-7. The organ- and cell-specific expression of this gene suggests that it may play a role in the regulation of cardiovascular and renal function, as well as fertility. In addition, the encoded protein is a functional receptor for the spike glycoprotein of the human coronaviruses SARS and HCoV-NL63.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse ACE2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse ACE2 standard solution and each sample is measured in duplicate.