Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse FGF1 ELISA Kit, Rockland Immunochemicals
Mouse FGF1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-830
CAS Number:  
Mouse FGF1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-830
Supplier Number:  KOA0675
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse FGF1 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-830

Specifications

About this item

Mouse FGF1 AccuSignal ELISA Kit

  • Natural and recombinant mouse FGF1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: F16-D155

Heparin-binding growth factor 1 is a protein that in humans is encoded by the FGF1 gene. The protein encoded by this gene is a member of the fibroblast growth factor(FGF) family. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis. The FGF1 gene was mapped to chromosome 5q31.3-q33.2 by in situ hybridization.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse FGF1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse FGF1 standard solution and each sample be measured in duplicate.