Order Entry
Puerto Rico
ContactUsLinkComponent
Human Nesfatin-1 ELISA Kit, Rockland Immunochemicals
Human Nesfatin-1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-818
CAS Number:  
Human Nesfatin-1 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-818
Supplier Number:  KOA0652
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Nesfatin-1 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-818

Specifications

About this item

Human Nesfatin-1 AccuSignal ELISA Kit

  • Natural and recombinant mouse CCL8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: V25-L106

Nesfatin-1 is a naturally occurring protein molecule produced by the brains of mammals. It is responsible for regulating appetite and production of body fat. Nesfatin-1 is a metabolic polypeptide encoded in the N-terminal region of the protein precursor, Nucleobindin2(NUCB2). Recombinant human Nesfatin-1 is a 9.7 kDa protein containing 82 amino acid residues. Originally identified as a hypothalamic neuropeptide, Nesfatin-1 is also expressed in other areas of the brain, and in pancreatic islets of Langerhans, gastric endocrine cells and adipocytes. Nesfatin-1 plays a role in hunger and energy regulation in an independent manner.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human Nesfatin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human Nesfatin-1 standard solution and each sample be measured in duplicate.
Seal the plate with the cover and incubate at 37°C for 90 min.