Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse CCL8 - MCP2 ELISA Kit, Rockland Immunochemicals
Mouse CCL8 - MCP2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-816
CAS Number:  
Mouse CCL8 - MCP2 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-816
Supplier Number:  KOA0651
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse CCL8 - MCP2 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-816

Specifications

About this item

Mouse CCL8 - MCP2 AccuSignal ELISA Kit

  • Natural and recombinant mouse CCL8.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: E20-P97

Chemokine(C-C motif) ligand 8, also known as monocyte chemoattractant protein 2(MCP-2), is a protein that in humans is encoded by the CCL8 gene. CCL8 is a small cytokine belonging to the CC chemokine family. The CCL8 protein is produced as a precursor containing 109 amino acids, which is cleaved to produce mature CCL8 containing 75 amino acids. The gene for CCL8 is encoded by 3 exons and is located within a large cluster of CC chemokines on chromosome 17q11.2 in humans. Monocyte chemotactic protein 2(MCP-2) is a CC chemokine that utilizes multiple cellular receptors to attract and activate human leukocytes.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse CCL8 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse CCL8 standard solution and each sample is measured in duplicate.