Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse GDF-15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-812
CAS Number:  
Mouse GDF-15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-812
Supplier Number:  KOA0649
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse GDF-15 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-812

Specifications

About this item

Mouse GDF-15 AccuSignal ELISA Kit

  • Natural and recombinant mouse GDF-15.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: S189-A303

GDF-15(Growth differentiation factor 15),also known as TGF-PL, MIC-1, PDF, PLAB, and PTGFB, is a protein belonging to the transforming growth factor beta superfamily that has a role in regulating inflammatory and apoptotic pathways in injured tissues and during disease processes. Using FISH, the MIC1 gene is mapped to 19p13.2-p13.1. Its expression in liver can be significantly up-regulated in during injury of organs such as liver, kidney, heart and lung. GDF15 showed increased expression and secretion during erythroblast maturation. GDF15 functions as an antiinflammatory cytokine by directly interfering with chemokine signaling and integrin activation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml mouse GDF-15 standard solutions into the pre-coated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse GDF-15 standard solution and each sample is measured in duplicate.