Order Entry
Puerto Rico
ContactUsLinkComponent
Human Galectin-9 ELISA Kit, Rockland Immunochemicals
Human Galectin-9 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-790
CAS Number:  
Human Galectin-9 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-790
Supplier Number:  KOA0634
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Galectin-9 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-790

Specifications

About this item

Human Galectin-9 AccuSignal ELISA Kit

  • Natural and recombinant human Galectin-9.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: M1-T355

Galectin-9, also called HUAT or LGALS9A is a protein that in humans is encoded by the LGALS9 gene. This gene is mapped to 17q11.2. The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. The protein encoded by this gene is an S-type lectin. It is overexpressed in Hodgkin's disease tissue and might participate in the interaction between the H&RS cells with their surrounding cells and might thus play a role in the pathogenesis of this disease and/or its associated immunodeficiency. The protein has N- and C- terminal carbohydrate-binding domains connected by a link peptide.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.7pg/m human Galectin-9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human Galectin-9 standard solution and each sample be measured in duplicate.