- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse TNFRSF4 - OX40 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-770
Mouse TNFRSF4 - OX40 AccuSignal ELISA Kit
- Natural and recombinant mouse TNFRSF4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: V20-P211
Tumor necrosis factor receptor superfamily, member4, also known as ACT35 or CD134 is a cell surface glycoprotein that was discovered through the production of a monoclonal antibody raised against the HUT-102 cell line. It belongs to the tumor necrosis factor receptor superfamily. CD134 was mapped to 1p36 by fluorescence in situ hybridization. CD134 is the primary receptor for feline immunodeficiency virus. CD134 expression can promote viral binding and renders cells permissive for viral entry, productive infection, and syncytium formation. Stimulating the receptor can improve the response to a powerful virus vector and may be useful in vaccine development.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse TNFRSF4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse TNFRSF4 standard solution and each sample be measured in duplicate.