Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Chitinase 3-like 1 - YKL-40 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-436
Specifications
About this item
Mouse Chitinase 3-like 1 - YKL-40 AccuSignal ELISA Kit
- Natural and recombinant mouse Chitinase 3-like 1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Y22-A381
Chitinase-3-like protein 1, also called CHI3L1 or YKL-40 is a secreted glycoprotein that is approximately 40kDa in size that in humans is encoded by the CHI3L1 gene. This gene is mapped to 1q32.1. Chitinases catalyze the hydrolysis of chitin, which is an abundant glycopolymer found in insect exoskeletons and fungal cell walls. The glycoside hydrolase 18 family of chitinases includes eight human family members. This gene encodes a glycoprotein member of the glycosyl hydrolase 18 family. The protein lacks chitinase activity and is secreted by activated macrophages, chondrocytes, neutrophils and synovial cells. The protein is thought to play a role in the process of inflammation and tissue remodeling.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse Chitinase 3-like 1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Chitinase 3-like 1 standard solution and each sample is measured in duplicate.