Order Entry
Puerto Rico
ContactUsLinkComponent
Human MICB ELISA Kit, Rockland Immunochemicals
Human MICB ELISA Kit, Rockland Immunochemicals
Catalog # 76235-724
CAS Number:  
Human MICB ELISA Kit, Rockland Immunochemicals
Catalog # 76235-724
Supplier Number:  KOA0593
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human MICB ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-724

Specifications

About this item

Human MICB AccuSignal ELISA Kit

  • Natural and recombinant human MICB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A23-G298

MHC class I polypeptide-related sequence B, also called MICB is a protein that in humans is encoded by the MICB gene. The MICB gene is mapped to 6p21.33. This gene encodes a heavily glycosylated protein which is a ligand for the NKG2D type II receptor. Binding of the ligand activates the cytolytic response of natural killer(NK) cells, CD8 alphabeta T cells, and gammadelta T cells which express the receptor. This protein is stress-induced and is similar to MHC class I molecules; however, it does not associate with beta-2-microglobulin or bind peptides. It seems to have no role in antigen presentation. This gene acts as a stress-induced self-antigen that is recognized by gamma delta T cells. It is the ligand for the KLRK1/NKG2D receptor and binds to KLRK1 leads to cell lysis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human MICB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human MICB standard solution and each sample be measured in duplicate.