Specifications
- Primary antibody reactivity:Human
- Target protein:ICAM-3
- Description:Human ICAM-3 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-690
Specifications
About this item
Human ICAM-3 AccuSignal ELISA Kit
- Natural and recombinant human ICAM-3.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q30-H485, homodimer
Intercellular adhesion molecule 3(ICAM3) also known as CD50(Cluster of Differentiation 50), is a human gene. The protein encoded by this gene is a member of the intercellular adhesion molecule(ICAM) family. ICAM3 is closely related to ICAM1, consists of 5 immunoglobulin domains, and binds LFA1 through its 2 N-terminal domains. This protein is constitutively and abundantly expressed by all leucocytes and may be the most important ligand for LFA-1 in the initiation of the immune response. It functions not only as an adhesion molecule, but also as a potent signalling molecule. ICAM3 has been shown to interact with EZR and Moesin.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human ICAM-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ICAM-3 standard solution and each sample be measured in duplicate.