- Primary antibody reactivity:Human
- Description:Human KIM1 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-660
Human KIM1 AccuSignal ELISA Kit
- Natural and recombinant human KIM1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S21-T288
KIM1(TIM-1), also known as Hepatitis A virus cellular receptor 1, is a protein that in Rats is encoded by the HAVCR1 gene. Infection of canine osteogenic sarcoma cells expressing HAVCR1 with HAV led to conclude that the protein is indeed a receptor for the virus. Immunofluorescence microscopy demonstrated internalization of HAV by dog cells expressing HAVCR1. Using a monoclonal antibody to Rat Tim1, Tim1 was expressed after activation of naive T cells and on T cells differentiated in Th2-polarizing conditions. By homology of synteny with the Rat Tim1 gene and database analysis, the HAVCR1 gene was mapped to 5q33.2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human KIM1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, cell lysates, serum, plasma(heparin, EDTA) or urine to each empty well. It is recommended that each human KIM1 standard solution and each sample be measured in duplicate.