Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse TrkB ELISA Kit, Rockland Immunochemicals
Mouse TrkB ELISA Kit, Rockland Immunochemicals
Catalog # 76235-640
CAS Number:  
Mouse TrkB ELISA Kit, Rockland Immunochemicals
Catalog # 76235-640
Supplier Number:  KOA0510
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse TrkB ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-640

Specifications

About this item

Mouse TrkB AccuSignal ELISA Kit

  • Natural and recombinant mouse TrkB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: C32-H429

TrkB receptor also known as TrkB tyrosine kinase or BDNF/NT-3 growth factors receptor or neurotrophic tyrosine kinase, receptor, type 2 is a protein that in humans is encoded by the NTRK2 gene.[1] TrkB is the high affinity catalytic receptor for several "neurotrophins", which are small protein growth factors that induce the survival and differentiation of distinct cell populations. The TrkB receptor is part of the large family of receptor tyrosine kinases. A "tyrosine kinase" is an enzyme which is capable of adding a phosphate group to certain tyrosines on target proteins, or "substrates".Soppet et al. [2] demonstrated that the gp145 gene product of the TRKB gene is rapidly phosphorylated on tyrosine residues upon exposure to BDNF and NTF3. The standard product used in this kit is recombinant TrkB, a 49.5KDa glycoprotein, C32—H439.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.8pg/ml mouse trkB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, cell lysates or tissue lysates to each empty well. We recommend that each mouse trkB standard solution and each sample is measured in duplicate.