- Primary antibody reactivity:Human
- Description:Human NOV - CCN3 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-360
Human NOV - CCN3 AccuSignal ELISA Kit
- Natural and recombinant human NOV.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: T32-M357
NOV(nephroblastoma overexpressed), also known as CCN3, is a matricellular protein that in humans is encoded by the NOV gene. The protein encoded by this gene is a small secreted cysteine-rich protein and a member of the CCN family of regulatory proteins. This gene is mapped to 8q24.12. NOV is a potentially useful marker for the diagnosis of adrenal gland diseases, malignant adrenocortical tumors, multiple sclerosis and so on. Moreover, reduced expression of NOV in ACTs may play an important role in the process of childhood ACT tumorigenesis. Though studying, it identified Nov as a regulator of human hematopoietic stem or progenitor cells.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human NOV standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human NOV standard solution and each sample be measured in duplicate.