- Primary antibody reactivity:Human
- Description:Human RANK ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-352
Human RANK AccuSignal ELISA Kit
- Natural and recombinant human RANK.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q29-G213
Receptor Activator of Nuclear Factor kappa B(RANK), also known as TRANCE Receptor, is a type I membrane protein that is expressed on the surface of osteoclasts and is involved in their activation upon ligand binding. RANK is a recently described TNF receptor family member, and its ligand, RANKL, promote survival of dendritic cells and differentiation of osteoclasts. RANK contains 383 amino acids in its intracellular domain(residues 234-616), which contain three putative TRAF-binding domains(termed I, II, and III). RANK interacts with various TRAFs through distinct motifs and activates NF-kappaB via a novel TRAF6 interaction motif, which then activates NIK, thus leading to NF-kappaB activation, whereas RANK most likely activates JNK through a TRAF2-interacting region in RANK. The standard in this kit is recombinant human RANK with the sequence of Q29-G213 aa. It is a dipolymer which compose of two chains, and the molecular weight of each is 48kda.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human RANK standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human RANK standard solution and each sample be measured in duplicate.