- Primary antibody reactivity:Human
- Description:Human Rage ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-348
Human Rage AccuSignal ELISA Kit
- Natural and recombinant human Rage.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q24-A344
RAGE, the Receptor for Advanced Glycation Endproducts, is a 35kD transmembrane receptor of the immunoglobulin super family. It is also known as “AGER”. AGER gene is mapped to chromosome 6p21.3 by mapping by contiguous cosmids and YAC clones and by fluorescence in situ hybridization. The expression of RAGE is particularly increased in neurons close to deposits of amyloid beta peptide and to neurofibrillary tangles. RAGE has been linked to several chronic diseases, which are thought to result from vascular damage. The pathogenesis is hypothesized to include ligand binding upon which RAGE signals activation of the nuclear factor kappa B(NF-kappaB).
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml, 78pg/ml human Rage standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. We recommend that each human Rage standard solution and each sample is measured in duplicate.