Order Entry
Puerto Rico
ContactUsLinkComponent
Human MSP - MST1 ELISA Kit, Rockland Immunochemicals
Human MSP - MST1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-914
CAS Number:  
Human MSP - MST1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-914
Supplier Number:  KOA0485
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human MSP - MST1 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-914

Specifications

About this item

Human MSP - MST1 AccuSignal ELISA Kit

  • Natural and recombinant human MSP.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: Q19-G711

Macrophage Stimulating 1(SMT1), also known as Hepatocyte growth factor-like protein(HGFL) and MSP, is a protein that in humans is encoded by the MST1 gene. The HGFL gene was identified at the DNF15S2 locus on human chromosome 3(3p21). The RON tyrosine kinase), the receptor for MSP, is expressed on the ciliated epithelia of the mucociliary transport apparatus of the lung. Furthermore, MSP stimulated ciliary motility in these cells by activating RON.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human MSP standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human MSP standard solution and each sample be measured in duplicate.