Order Entry
Puerto Rico
ContactUsLinkComponent
Human CSF1R - M-CSFR ELISA Kit, Rockland Immunochemicals
Human CSF1R - M-CSFR ELISA Kit, Rockland Immunochemicals
Catalog # 76236-900
CAS Number:  
Human CSF1R - M-CSFR ELISA Kit, Rockland Immunochemicals
Catalog # 76236-900
Supplier Number:  KOA0478
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CSF1R - M-CSFR ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-900

Specifications

About this item

Human CSF1R - M-CSFR AccuSignal ELISA Kit

  • Natural and recombinant human CSF1R.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: I20-E512

CSF1R(Colony stimulating factor 1 receptor),also known as M-CSFR and CD115, is a cell-surface protein encoded, in humans, by the CSF1R gene. The gene is located on long arm of chromosome 5(5q32) on the Crick(minus) strand. The encoded protein is a tyrosine kinase transmembrane receptor and member of the CSF1/PDGF receptor family of tyrosine-protein kinases. The encoded protein is a single pass type I membrane protein and acts as the receptor for colony stimulating factor 1, a cytokine which controls the production, differentiation, and function of macrophages. Both CSF1R, and its ligand colony stimulating factor 1 play an important role in the development of the mammary gland and may be involved in the process of mammary gland carcinogenesis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human CSF1R standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CSF1R standard solution and each sample be measured in duplicate.