- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse MBL2 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-338
Mouse MBL2 AccuSignal ELISA Kit
- Natural and recombinant mouse MBL2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E19-D244
MBL2, also called mannose-binding lectin (protein C) 2, soluble or Mannose-binding lectin (MBL) is a lectin that is instrumental in innate immunity. MBL2 is mapped to chromosome 10q11.2-q21. It belongs to the class of collectins in the C-type lectin superfamily, whose function appears to be pattern recognition in the first line of defense in the pre-immune host. MBL2 recognizes carbohydrate patterns, found on the surface of a large number of pathogenic micro-organisms, including bacteria, viruses, protozoa and fungi. Binding MBL2 to a micro-organism results in activation of the lectin pathway of the complement system. Another important function of MBL2 is that this molecule binds senescent and apoptotic cells and enhances engulfment of whole, intact apoptotic cells, as well as cell debris by phagocytes.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse MBL2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse MBL2 standard solution and each sample be measured in duplicate.