Order Entry
Puerto Rico
ContactUsLinkComponent
 
Human XCL1 - Lymphotactin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-332
CAS Number:  
Human XCL1 - Lymphotactin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-332
Supplier Number:  KOA0474
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human XCL1 - Lymphotactin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-332

Specifications

About this item

Human XCL1 - Lymphotactin AccuSignal ELISA Kit

  • Natural and recombinant human XCL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: V22-G114

Chemokine(C motif) ligand(XCL1) is a small cytokine belonging to the XC chemokine family that is also known as lymphotactin. This gene is mapped to chromosome 1q23 by FISH. The sequence of the deduced 114-amino acid protein is most homologous to the CC chemokines CCL8 and CCL3. It is found in high levels in spleen, thymus, intestine and peripheral blood leukocytes, and at lower levels in lung, prostate gland and ovary. XCL1 induces the migration of cells expressing XCR1.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human XCL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each human XCL1 standard solution and each sample be measured in duplicate.