Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse IL-21 ELISA Kit, Rockland Immunochemicals
Mouse IL-21 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-326
CAS Number:  
Mouse IL-21 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-326
Supplier Number:  KOA0470
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-21
  • Description:
    Mouse IL-21 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-326

Specifications

About this item

Mouse Interleukin-21 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-21.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.colI; Immunogen sequence: P25-S146

Interleukin-21 is a protein that in humans is encoded by the IL21 gene. Interleukin 21 is a cytokine that has potent regulatory effects on cells of the immune system, including natural killer(NK) cells and cytotoxic T cells that can destroy virally infected or cancerous cells. By radiation hybrid analysis, the IL21 gene is mapped to chromosome 4q26-q27, near the IL2 gene. IL21 stimulates B-cell proliferation in an anti-CD40-dependent manner but inhibits proliferation stimulated by IL4 plus anti-IgM. The biologic effects of IL21 on NK cells differ between species, with human IL21 enhancing NK cell proliferation and murine IL21 inhibiting NK cell proliferation. IL21 is a cytokine highly expressed by mouse T(H)17 cells.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse IL-21 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse IL-21 standard solution and each sample be measured in duplicate.