Order Entry
Puerto Rico
ContactUsLinkComponent
Human IL-17RB ELISA Kit, Rockland Immunochemicals
Human IL-17RB ELISA Kit, Rockland Immunochemicals
Catalog # 76237-314
CAS Number:  
Human IL-17RB ELISA Kit, Rockland Immunochemicals
Catalog # 76237-314
Supplier Number:  KOA0464
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-17RB
  • Description:
    Human IL-17RB ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-314

Specifications

About this item

Human Interleukin-17RB AccuSignal ELISA Kit

  • Natural and recombinant human IL-17RB.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NAO; Immunogen sequence: R18-K286

IL17RB, also known as CRL4, is a protein that in humans is encoded by the IL17RB gene. It is located on 3p21.1. IL17RB is produced by activated T cells exhibiting proinflammatory activities. The protein encoded by this gene is a cytokine receptor. This receptor specifically binds to IL17B and IL17E (IL25), but does not bind to IL17 (A) or IL17C. This receptor has been shown to mediate the activation of NF-kappaB and the production of IL8 induced by IL17E. And it may play a role in controlling the growth and/or differentiation of hematopoietic cells.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human IL-17RB standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IL-17RB standard solution and each sample be measured in duplicate.