Order Entry
Puerto Rico
ContactUsLinkComponent
Human IGF1R ELISA Kit, Rockland Immunochemicals
Human IGF1R ELISA Kit, Rockland Immunochemicals
Catalog # 76235-632
CAS Number:  
Human IGF1R ELISA Kit, Rockland Immunochemicals
Catalog # 76235-632
Supplier Number:  KOA0457
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human IGF1R ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-632

Specifications

About this item

Human IGF1R AccuSignal ELISA Kit

  • Natural and recombinant human IGF1R.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E31-N932

IGF1R, also called IGFR or insulin-like growth factor 1 receptor, is a protein that found on the surface of human cells. This gene is mapped to 15q26.3. IGF1R belongs to the large class of tyrosine kinase receptors. This receptor mediates the effects of IGF-1, which is a polypeptide protein hormone similar in molecular structure to insulin. It plays an important role in growth and continues to have anabolic effects in adults - meaning that it can induce hypertrophy of skeletal muscle and other target tissues. IGF1R also plays a critical role in transformation events. It is highly overexpressed in most malignant tissues where it functions as an anti-apoptotic agent by enhancing cell survival.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human IGF1R standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IGF1R standard solution and each sample be measured in duplicate.