Order Entry
Puerto Rico
ContactUsLinkComponent
Human GDF-15 ELISA Kit, Rockland Immunochemicals
Human GDF-15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-624
CAS Number:  
Human GDF-15 ELISA Kit, Rockland Immunochemicals
Catalog # 76235-624
Supplier Number:  KOA0453
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human GDF-15 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76235-624

Specifications

About this item

Human GDF-15 AccuSignal ELISA Kit

  • Natural and recombinant human GDF-15.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: A197-I308

GDF-15(Growth differentiation factor 15),also known as TGF-PL, MIC-1, PDF, PLAB, and PTGFB, is a protein belonging to the transforming growth factor beta superfamily that has a role in regulating inflammatory and apoptotic pathways in injured tissues and during disease processes. Using FISH, the MIC1 gene is mapped to 19p13.2-p13.1. Its expression in liver can be significantly up-regulated in during injury of organs such as liver, kidney, heart and lung. GDF15 showed increased expression and secretion during erythroblast maturation. GDF15 functions as an antiinflammatory cytokine by directly interfering with chemokine signaling and integrin activation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml human GDF-15 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human GDF-15 standard solution and each sample be measured in duplicate.