- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Galectin-1 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-616
Mouse Galectin-1 AccuSignal ELISA Kit
- Natural and recombinant mouse Galectin-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.colI; Immunogen sequence: A2-E135
Galectin-1 is a protein that in humans is encoded by the LGALS1 gene.The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. LGALS1 may act as an autocrine negative growth factor that regulates cell proliferation.Baldini et al. stated that the mouse beta-galactoside-binding protein is an autocrine regulator of cell proliferation with a role in the maintenance of G0 and in the control of G2 traverse. They found that galectin-1 was expressed in a subset of slowly dividing subventricular zone astrocytes, which included the neural stem cells. Intraventricular infusion experiments and phenotypic analysis of knockout mice showed that galectin-1 was an endogenous factor that promoted the proliferation of neural stem cells in adult mouse brain. The standard product used in this kit is recombinant mouse GALECTIN-1, Ala2-Glu135, with the molecular mass of 15KDa.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse Galectin-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse Galectin-1 standard solution and each sample be measured in duplicate.