Order Entry
Puerto Rico
ContactUsLinkComponent
Rat CXCL2 - MIP-2 ELISA Kit, Rockland Immunochemicals
Rat CXCL2 - MIP-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-286
CAS Number:  
Rat CXCL2 - MIP-2 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-286
Supplier Number:  KOA0422
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Rat CXCL2 - MIP-2 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-286

Specifications

About this item

Rat CXCL2 - MIP-2 AccuSignal ELISA Kit

  • Natural and recombinant rat MIP-2.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S32-N100

MIP is a member of the aquaporin family of membrane-bound water channels. MIP family proteins are thought to contain 6 TM domains. Sequence analysis suggests that the proteins may have arisen through tandem, intragenic duplication from an ancestral protein that contained 3 TM domains. Major intrinsic protein (MIP, also called MP26) is the predominant fiber cell membrane protein of the ocular lens. The major intrinsic protein (MIP) of the vertebrate eye lens is the first identified member of a sequence-related family of cell-membrane proteins that appears to have evolved by gene duplication. Several members of the MIP family transport water (aquaporins), glycerol and other small molecules in microbial, plant and animal cells.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat MIP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat MIP-2 standard solution and each sample be measured in duplicate.