Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Adiponectin ELISA Kit, Rockland Immunochemicals
Mouse Adiponectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-198
CAS Number:  
Mouse Adiponectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-198
Supplier Number:  KOA0366
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Adiponectin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-198

Specifications

About this item

Mouse Adiponectin AccuSignal ELISA Kit

  • Natural and recombinant full length mouse Adiponectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: E18-N247

Adiponectin(ADPN) is a hormone secreted by adipocytes that regulates energy homeostasis and glucose and lipid metabolism. Adiponectin is a new member of the family of soluble defense collagens, in hematopoiesis and immune responses. It is an important negative regulator in hematopoiesis and immune systems and raise the possibility that it may be involved in ending inflammatory responses through its inhibitory functions. Adiponectin is mapped to 3q27 and can protect the organism from systemic inflammation by promoting the clearance of early apoptotic cells by macrophages through a receptor-dependent pathway involving calreticulin.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20000pg/ml, 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml mouse Adiponectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum, plasma(heparin, EDTA), tissue homogenates or urine to each empty well. It is recommended that each mouse Adiponectin standard solution and each sample be measured in duplicate.