Order Entry
Puerto Rico
ContactUsLinkComponent
Human VEGF-C ELISA Kit, Rockland Immunochemicals
Human VEGF-C ELISA Kit, Rockland Immunochemicals
Catalog # 76236-864
CAS Number:  
Human VEGF-C ELISA Kit, Rockland Immunochemicals
Catalog # 76236-864
Supplier Number:  KOA0359
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Target protein:
    VEGF-C
  • Description:
    Human VEGF-C ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-864

Specifications

About this item

Human VEGF-C AccuSignal ELISA Kit

  • Natural human VEGF-C, recombinant human VEGF-C (wild type), and recombinant human VEGF-C (Cys156Ser substituted).
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: T103-R227

Vascular endothelial growth factor C is a VEGF. The human gene encoding it is VEGFC. The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor(PDGF/VEGF) family, is active in angiogenesis, lymphangiogenesis and endothelial cell growth and survival, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D(VEGF-D). The C terminus of VEGFC has cysteine-rich repeat units characteristic of the Balbiani ring 3 protein(BR3P) of the midge Chironomus tentans.1,2 The standard product used in this kit is recombinant human VEGF-C, consisting of 135 amino acids with the molecular mass of 23Kda after glycosylation.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml,2000pg/ml, 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml human VEGF-C standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human VEGF-C standard solution and each sample be measured in duplicate.