- Primary antibody reactivity:Mouse
- Target protein:MCP-1 (CCL2)
- Description:Mouse MCP-1 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-596
Mouse MCP-1 AccuSignal ELISA Kit
- Natural and recombinant mouse MCP-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: Q24-R96
Monocyte chemotactic protein-1(MCP-1) is a member of the small inducible gene(SIG) family. It has been shown to play a role in the recruitment of monocytes to sites of injury and infection. By analysis of a panel of somatic cell hybrids, we have localized the MCP-1 gene, designated SCYA2, to human chromosome 17. In situ hybridization confirmed this assignment and further localized the gene to 17q11.2-q21.1. MCP-1 plays a unique and crucial role in the initiation of atherosclerosis and may provide a new therapeutic target in this disorder. Monocyte chemoattractant protein-1(MCP-1), regulated on activation, normal T cell expressed and secreted,and stimulation of monocytes from healthy carriers of the genotype GG with Mycobacterium tuberculosis antigens yielded higher MCP-1. The standard used in this kit is recombinant mouse MCP-1(Q24-R96) with the molecular mass of 8.5Kda.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse MCP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse MCP-1 standard solution and each sample be measured in duplicate.