Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse CCL1 - TCA3 ELISA Kit, Rockland Immunochemicals
Mouse CCL1 - TCA3 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-850
CAS Number:  
Mouse CCL1 - TCA3 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-850
Supplier Number:  KOA0344
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse CCL1 - TCA3 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-850

Specifications

About this item

Mouse CCL1 - TCA3 AccuSignal ELISA Kit

  • Natural and recombinant mouse CCL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: K24-C92

CCL1, Chemokine(C-C motif) ligand 1, is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml mouse CCL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse CCL1 standard solution and each sample is measured in duplicate.