Order Entry
Puerto Rico
ContactUsLinkComponent
Human CCL1 ELISA Kit, Rockland Immunochemicals
Human CCL1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-848
CAS Number:  
Human CCL1 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-848
Supplier Number:  KOA0343
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human CCL1 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-848

Specifications

About this item

Human CCL1 AccuSignal ELISA Kit

  • Natural and recombinant human CCL1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: K24-K96

CCL1, Chemokine(C-C motif) ligand 1, is one of several cytokine genes clustered on the q-arm of chromosome 17. Cytokines are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The protein encoded by this gene is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine is secreted by activated T cells and displays chemotactic activity for monocytes but not for neutrophils. It binds to the chemokine receptor CCR8.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human CCL1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CCL1 standard solution and each sample be measured in duplicate.