- Primary antibody reactivity:Human
- Description:Human Cardiotrophin-1 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-846
Human Cardiotrophin-1 AccuSignal ELISA Kit
- Natural and recombinant human CT-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S2-A201
Cardiotrophin-1(CT-1) is a member of the family of cytokines that includes leukemia inhibitory factor(LIF), ciliary neurotrophic factor(CNTF), oncostatin M(OSM), interleukin-6(IL6), and interleukin-11(IL11). And the CT-1 gene is mapped to 1p21-p13. The human CT-1 protein contains 201 amino acids and shares 80% amino acid identity with the 203-amino acid mouse CT-1 sequence; however, unlike the mouse protein, human CT-1 has 2 rather than 1 cys and has no N-glycosylation site. Despite lacking a signal sequence, secreted CT-1 and mouse CT-1 induce cardiac myocyte hypertrophy in cell culture and bind to both mouse and human LIFR but not to OSMR. Furthermore, A 1.7-kb CT-1 transcript was detected at high levels in heart, skeletal muscle, prostate, and ovary. Low levels were detected in lung, kidney, pancreas, thymus, testis, and small intestine, with little or no expression detected in brain, placenta, spleen, colon, and peripheral blood leukocytes. And it was also observed strong expression in fetal lung and kidn...
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human CT-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human CT-1 standard solution and each sample be measured in duplicate.