Order Entry
Puerto Rico
ContactUsLinkComponent
Rat TNF alpha ELISA Kit, Rockland Immunochemicals
Rat TNF alpha ELISA Kit, Rockland Immunochemicals
Catalog # 76236-798
CAS Number:  
Rat TNF alpha ELISA Kit, Rockland Immunochemicals
Catalog # 76236-798
Supplier Number:  KOA0318
CAS Number:  

Specifications

  • Target protein:
    TNF-alpha
  • Description:
    Rat TNF alpha ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-798

Specifications

About this item

Rat TNF alpha AccuSignal ELISA Kit

  • Natural and recombinant rat TNF alpha.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: L80-L235

Tumor necrosis factor-alpha(TNF-alpha, or TNF) is secreted by macrophages in response to inflammation, infection and cancer. Human Tumor Necrosis Factor(TNF) and Lymphotoxin(TNF-beta) are cytotoxic proteins which have similar biological activities and share 30% amino acid homology. TNF-alpha is produced by monocytes, which can stimulate endothelial cells to produce the multilineage growth factor granulocyte-macrophage colony-stimulating factor and extend the role of this immunoregulatory protein to the regulation of hematopoiesis in vitro. TNF is a soluble protein that causes damage to tumor cells but has no effect on normal cells. Human TNF has been purified to apparent homogeneity as a 17.3-kilodalton protein from HL-60 leukemia cells and has showed cytotoxic and cytostatic activities against various human tumor cell lines. The human TNF cDNA is 1585 base pairs in length and encodes a protein of 233 amino acids. The mature protein begins at residue 77, leaving a long leader sequence of 76 amino acids. TNF-a...

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat TNF alpha standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin , EDTA) to each empty well. It is recommended that each rat TNF alpha standard solution and each sample be measured in duplicate.