- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Human
- Target protein:TIMP-2
- Description:Human TIMP-2 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-790
Human TIMP-2 AccuSignal ELISA Kit
- Natural and recombinant human TIMP-2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: CHO; Immunogen sequence: C27-P220
TIMP2 gen is encoded by 5 exons spanning 83 kb of genomic DNA. TIMP2 is 83 kilobase pairs(kb) long with exon-intron splicing sites located in preserved positions among the three members of the TIMP family. The gene for tissue inhibitor of metalloproteinases-2 is localized on human chromosome arm 17q25. TIMP-2 abrogates angiogenic factor-induced endothelial cell proliferation in vitro and angiogenesis in vivo independent of MMP inhibition. The standard product used in this kit is recombinant human TIMP-2 with the molecular mass of 22Kda and 194 amino acid.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TIMP-2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin , EDTA) or saliva to each empty well. It is recommended that each human TIMP-2 standard solution and each sample be measured in duplicate.