Order Entry
Puerto Rico
ContactUsLinkComponent
Rat TGF beta 1 ELISA Kit, Rockland Immunochemicals
Rat TGF beta 1 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-194
CAS Number:  
Rat TGF beta 1 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-194
Supplier Number:  KOA0308
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Rat
  • Target protein:
    TGF beta 1
  • Description:
    Rat TGF beta 1 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-194

Specifications

About this item

Rat TGF beta 1 AccuSignal ELISA Kit

  • Natural and recombinant rat TGF beta 1.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: CHO; Immunogen sequence: A279-S390

Transforming growth factor-beta1(TGF-beta1) is a multifunctional peptide that controls proliferation, differentiation, and other functions in many cell types. Many cells synthesize TGF-beta and essentially all of them have specific receptors for this peptide. TGF-beta regulates the actions of many other peptide growth factors and determines a positive or negative direction of their effects. TGFbeta1 is known for its potent and diverse biological effects, including immune regulation, and cell growth and differentiation. TGFbeta1 is also an important mediator of bone remodeling. TGFbeta1, a potent keratinocyte growth inhibitor, has been shown to be overexpressed in keratinocytes in certain inflammatory skin diseases and has been thought to counteract the effects of other growth factors at the site of inflammation. TGF-beta1, a multifunctional cytokine with fibrogenic properties, has been implicated in the pathogenesis of the vascular and target organ complications of hypertension. TGF-beta1 may also regulate bl...

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat TGF beta 1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum, plasma (EDTA) or urine to each empty well. It is recommended that each rat TGF beta 1 standard solution and each sample be measured in duplicate.