Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse L-Selectin ELISA Kit, Rockland Immunochemicals
Mouse L-Selectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-178
CAS Number:  
Mouse L-Selectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-178
Supplier Number:  KOA0300
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse L-Selectin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-178

Specifications

About this item

Mouse L-Selectin AccuSignal ELISA Kit

  • Natural and recombinant mouse sL-Selectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: W39-N332

L-selectin, also known as CD62L, is a cell adhesion molecule found on leukocytes. It belongs to the selectin family of proteins, which recognize sialylated carbohydrate groups. It is cleaved by ADAM17.SELL(L-selectin)is a cell surface component that is a member of a family of adhesion/homing receptors which play important roles in leukocyte-endothelial cell interactions. The molecule is composed of multiple domains: one homologous to lectins, one to epidermal growth factor, and two to the consensus repeat units found in C3/C4 binding proteins.1L-selectin acts as a "homing receptor" for leukocytes to enter secondary lymphoid tissues via high endothelial venules. Ligands present on endothelial cells will bind to leukocyte expressing L-selectin, slowing leukocyte trafficking through the blood, and facilitating entry into a secondary lymphoid organ at that point2.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 4000pg/ml, 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml mouse L-Selectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin , EDTA) to each empty well. It is recommended that each mouse L-Selectin standard solution and each sample be measured in duplicate.