Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Neurotrophin-3 ELISA Kit, Rockland Immunochemicals
Rat Neurotrophin-3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-590
CAS Number:  
Rat Neurotrophin-3 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-590
Supplier Number:  KOA0281
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Rat Neurotrophin-3 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-590

Specifications

About this item

Rat Neurotrophin-3 AccuSignal ELISA Kit

  • Natural and recombinant rat Neurotrophin-3.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: Y139-T257

Neurotrophin-3(NT-3) is a new member of the nerve growth factor gene family, which plays an important role in the development and maintenance of the vertebrate nervous system. NT-3 and its receptor, called neurotrophic tyrosine kinase receptor type 3(Ntrk3; also called TrkC), are expressed early and throughout embryogenesis. NT-3 is one of five neurotrophin growth factors which shape the development of the nervous system by regulating neuronal survival and differentiation. NT-3 may be one of the central nervous system-derived factors that mediate neural crest(NC) cell proliferation in vivo. NT-3 has been mapped to human chromosome 12p and mouse chromosome 6.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat Neurotrophin-3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernatants or serum to each empty well. It is recommended that each rat Neurotrophin-3 standard solution and each sample be measured in duplicate.