Order Entry
Puerto Rico
ContactUsLinkComponent
Rat NGF - NGF beta ELISA Kit, Rockland Immunochemicals
Rat NGF - NGF beta ELISA Kit, Rockland Immunochemicals
Catalog # 76236-760
CAS Number:  
Rat NGF - NGF beta ELISA Kit, Rockland Immunochemicals
Catalog # 76236-760
Supplier Number:  KOA0278
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Rat NGF - NGF beta ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-760

Specifications

About this item

Rat NGF - NGF beta AccuSignal ELISA Kit

  • Natural and recombinant rat NGF.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: S122-G241

Nerve growth factor(NGF) is a polypeptide involved in the regulation of growth and differentiation of sympathetic and certain sensory neurons. NGF is thought to have a profound effect on the development and maintenance of sympathetic and embryonic sensory neurones. NGF activity isolated from the male mouse submaxillary gland(MSG) consists of three types of subunits, alpha, beta and gamma, which specifically interact to form a 7S, approximately 130,000-molecular weight(Mr) complex. The 7S complex contains two identical 118-amino acid beta-chains, which are solely responsible for the nerve growth-stimulating activity of NGF. NGF, which is expressed by inflammatory cells and effects changes that lead to increased neural responsiveness, could be a pivotal mediator in allergic rhinitis.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml rat NGF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernatants or serum to each empty well. It is recommended that each rat NGF standard solution and each sample be measured in duplicate.