- Primary antibody reactivity:Mouse
- Target protein:MMP-9
- Description:Mouse MMP-9 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-752
Mouse MMP-9 AccuSignal ELISA Kit
- Natural and recombinant mouse total MMP-9.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: CHO; Immunogen sequence: A20-P730
The 92-kD type IV collagenase is also known as 92-kD gelatinase, type V collagenase, gelatinase B, or matrix metalloproteinase-9(MMP9). The 72- and 92-kDa type IV collagenases are members of a group of secreted zinc metalloproteases.1 The matrix metalloproteinases(MMPs) are able to degrade the extracellular matrix and allow angiogenesis and tumor invasion. Gelatinase B, a matrix metalloproteinase that has proteolytic activity against connective tissue proteins, has been suggested to be important in the connective tissue remodeling processes associated with atherogenesis and plaque rupture. MMP-9 is predominantly expressed in neutrophils, macrophages, and mast cells, rather than in oncogene-positive neoplastic cells. The polymorphism of MMP-9 acts as a genetic factor for the development of smoking-induced pulmonary emphysema.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse MMP-9 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma( heparin) to each empty well. It is recommended that each mouse MMP-9 standard solution and each sample be measured in duplicate.