- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse CCL3 - MIP1 alpha ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-160
Mouse CCL3 - MIP1 alpha AccuSignal ELISA Kit
- Natural and recombinant mouse CCL3.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: A24-A92
Chemokine(C-C motif) ligand 3(CCL3), so known as Macrophage inflammatory protein-1alpha(MIP-1 alpha), is a protein that in humans is encoded by the CCL3 gene. It is a cytokine which belongs to the CC chemokine family that is involved in the acute inflammatory state in the recruitment and activation of polymorphonuclear leukocytes. This gene is mapped to 17q12. CCL3 plays a role in inflammatory responses through binding to the receptors CCR1, CCR4 and CCR5. It has been found that the activation of CCR5 by CCL3 directly and independently activates a G-protein signaling pathway through GNAI2 and a tyrosine phosphorylation signaling pathway through JAK2.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml mouse CCL3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. We recommend that each mouse CCL3 standard solution and each sample is measured in duplicate.