Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse M-CSF ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-720
Specifications
About this item
Mouse M-CSF AccuSignal ELISA Kit
- Natural and recombinant mouse M-CSF.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: K33-E262
M-CSF, also called CSF1, consists of a 14-amino acid peptide, longer than the usual 8-to-11 mer recognized by most CTLs. The M-CSF gene is mapped to 1p21-p13 and contains 10 exons and 9 introns spanning 20 kb1. Although it is a single-copy gene, its expression results in the synthesis of several mRNAs, ranging in size from about 1.5 to 4.5 kb2. There are 2 forms of M-CSF, with 224 and 522 amino acids, resulting from alternative splicing3. Furthermore, M-CSF has a role in development of the placenta. Uterine M-CSF concentration is regulated by a synergistic action of estradiol and progesterone. M-CSF is produced by uterine glandular epithelial cells. It had been found that FMS, the M-CSF receptor, is expressed in placenta and choriocarcinoma cell lines4.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse M-CSF standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, tissue lysates, serum or plasma (EDTA) to each empty well. It is recommended that each mouse M-CSF standard solution and each sample be measured in duplicate.