Order Entry
Puerto Rico
ContactUsLinkComponent
 
Mouse IL-5 ELISA Kit, Rockland Immunochemicals
  76236-672
 :  
Mouse IL-5 ELISA Kit, Rockland Immunochemicals
  76236-672
 :  KOA0224
 :  

 

  • Primary antibody reactivity:
    Mouse
  • Target protein:
    IL-5
  • Description:
    Mouse IL-5 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-672

 

 

Mouse Interleukin-5 AccuSignal ELISA Kit

  • Natural and recombinant mouse IL-5.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: SF21; Immunogen sequence: M21-G133

Interleukin-5(IL-5) is also known as eosinophil differentiation factor(EDF). IL-5 is a potential candidate gene in the pathogenesis of asthma, as it is the main cytokine controlling eosinophil activity and eosinophils are pivotal in the development of airway inflammation. The predicted amino acid sequence of 134 amino acids is identical with that recently reported for human interleukin-5 but shows no significant homology with other known hemopoietic growth regulators. IL-5 is a lineage-specific hematopoietic growth factor that stimulates the production of eosinophils and eosinophil colonies from normal human bone marrow cells. IL-5 gene to human chromosome 5 at bands q23-31. Human and mouse of IL-5 share 70% amino acid sequence homology.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse IL-5 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates or serum to each empty well. It is recommended that each mouse IL-5 standard solution and each sample be measured in duplicate.