Order Entry
Puerto Rico
ContactUsLinkComponent
Human Flt-3ligand ELISA Kit, Rockland Immunochemicals
Human Flt-3ligand ELISA Kit, Rockland Immunochemicals
Catalog # 76236-612
CAS Number:  
Human Flt-3ligand ELISA Kit, Rockland Immunochemicals
Catalog # 76236-612
Supplier Number:  KOA0172
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human Flt-3ligand ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-612

Specifications

About this item

Human Flt-3ligand AccuSignal ELISA Kit

  • Natural and recombinant human Flt-3ligand.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: sf21; Immunogen sequence: T27-P185

Fms-related tyrosine kinase 3 ligand(FLT3LG), also known as Flt-3ligand or FL, is a protein which in humans is encoded by the FLT3LG gene. It is mapped to 19q13.3. Flt-3ligand is a hematopoietic four helical bundle cytokine. It is structurally homologous to stem cell factor(SCF) and colony stimulating factor 1(CSF-1). In synergy with other growth factors, Flt-3ligand stimulates the proliferation and differentiation of various blood cell progenitors. Besides that, Flt-3ligand can control the development of Dendritic cells(DCs) and is particularly important for plasmacytoid DCs and CD8-positive classical DCs and their CD103-positive tissue counterparts. Flt-3ligand also can enhance the response of stem and primitive progenitor cells to other growth factors to generate all myeloid lineages except erythroid cells.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml human Flt-3ligand standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA, citrate) to each empty well. It is recommended that each human Flt-3ligand standard solution and each sample be measured in duplicate.