To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Oxphos™ Cell Survival Assay Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:75930-664
- Supplier no.:KLD-002
Specifications
About this item
Oxphos™ Cell Survival Assay Kit
- Oxphos™ contains enough reagents for 100 assays using a a 6-well plate.
OxPhos™ is a medium based metabolic probe that can measure glutathione recycling capacity/glutathione function and glucose dependent antioxidant capacity of cells in tissue culture and whole blood. Oxphos™ is based on the ability of mammalian cells to rapidly and efficiently convert hydroxyethyl disulfide (HEDS) into mercaptoethanol (ME) through a bioreduction mechanism. Bioconversion of HEDS to ME relies on the activity of the oxidative pentose phosphate cycle (OPPC). OxPhos™ measures ME in the extracellular medium without the need for cellular lysis and extraction methods. Cell media is used in the assay, avoiding the need to lyse cells and thereby saving time and cost while preserving the ability to perform other cellular tests in the same culture system. OxPhos™ may have multiple applications in aging, oxidative stress, antioxidant screening, chemotherapy response and toxicology.
Oxphos™ gives a linear response for mammalian cells (0; 100,000; 200,000; 400,000; 600,000; and 800,000) plated in 1mL growth medium in a six well plate with up to 15% fetal bovine serum and measured 20 hours after plating. It also gives a linear response for human blood (0, 10, 20, 40, 60, 80, 100 µL) suspended in a total volume of 200 µL saline in a microfuge tube and incubated with 10 µL of reagent 6 for 2hrs. Oxphos™ contains enough reagents for 100 assays using a a 6-well plate.