Specifications
- Description:CellCountEZ™ Cell Survival Assay Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:10800-766
- Supplier no.:KLD-001
Specifications
About this item
CellCountEZ™ Cell Survival Assay Kit
- This kits provides for 1000 assays using a 96-well plate.
CellCountEZ™ Cell Survival Assay Kit is a tissue culture media-based assay that can measure metabolically active live cells and quantify cell death caused by radiation, chemotherapeutics or toxins. CellCountEZ™ is based on the ability of mammalian cells to rapidly and efficiently convert hydroxyethyl disulfide (HEDS) into mercaptoethanol (ME) through a bioreduction mechanism. Bioconversion of HEDS to ME relies on the activity of the oxidative pentose phosphate cycle (OPPC). CellCountEZ™ measures ME in the extracellular medium without the need for cellular lysis and extraction methods, this preserves the ability to perform other cellular tests in the same culture system. CellCountEZ™ is readily soluble, membrane permeable and converted by live cells intracellularly before transport into the extracellular culture media. CellCountEZ™ offers many advantages that make it superior to common existing methods for quantifying cell growth and survival.
CellCountEZ™ Cell Survival Assay Kit contains enough reagents for 1000 assays using a 96-well plate. This assay gives a linear response for cells (0; 1,000; 5,000; 10,000; 20,000; 40,000) plated in 100 µl growth medium with up to 15% fetal bovine serum in a 96-well plate and measured 20 hours after plating. Drug toxicity could be measured for up to 4 days after treatment with drugs using this assay for 5,000 cells plated in 100 µl growth medium with up to 15% fetal bovine serum in a 96-well plate.