Lysis Buffer, 1X, Rockland Immunochemicals
1X Lysis Buffer, 50 mL
Catalog # 10799-486
Supplier: Rockland Immunochemical
: KBD-001
Lysis Buffer, 1X, Rockland Immunochemicals
Catalog # 10799-486
Supplier: Rockland Immunochemical
CAS Number:
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
1X Lysis Buffer
- 50 mM Tris Cl, 150 mM Sodium Chloride, 1 mM EDTA, 1% TRITON-X-100, pH 7.4.
- No further preparation is required prior to use. Chill to 2 to 8 °C prior to use.
Lysis Buffer is used to lyse cells under nondenaturing conditions. Cell Lysis Buffer is ideal to assist in Immunoprecipitation which allows for the purification by immunoprecipitation of recombinant proteins containing an epitope tag provided by the user. The user is able to further characterize the resultant purified protein by size, post-translational modification, western blot and other assays.
Chill to 2 to 8 °C prior to use. Lysis Buffer is provided as a ready-to-use 1X solution containing 50 mM Tris Cl, 150 mM Sodium Chloride, 1 mM EDTA, 1% TRITON-X-100, pH 7.4. No preparation is required prior to use.
Specifications
Specifications
- Description:1X Lysis Buffer
- Size:50 mL
- Cat. No.:10799-486
- Shipping Temperature:Wet Ice



